Thermo Scientific 900002 Inner chamber filling solution
Representative image only.
Thermo Scientific 900002 Inner chamber filling solution
Thermo Scientific | Mfr: 900002| Item: 0573039
Have refill solutions handy and minimize downtime
₹27,400.00INR / Carton of 5
₹27,400.00INR / Carton of 5
Product Description
Key Features
  • Five 60 mL bottles
More About this Item
Ensure accurate pH readings with reference fill solutions.
Specifications
  • Compatible WithReference electrode 05710-10
  • TypeInner fill solution
  • DescriptionpH Electrode Reference Inner Filling Solution, 5 x 60 mL