Thermo Scientific 941707 Standardizing solution: NaCl as Cl, 100 ppm
Representative image only.
Thermo Scientific 941707 Standardizing solution: NaCl as Cl, 100 ppm
Thermo Scientific | Mfr: 941707| Item: 0573019
Standardizing solutions calibrate your instrument
₹20,900.00INR / Each
₹20,900.00INR / Each
Product Description
Key Features
  • Help perform addition and subtraction operations
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Solutions work best when combined with ionic strength and pH adjustors, which eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. All standardizing solutions and ionic strength adjustors come in 475-ml bottles except model 05730-15, (1 gal), and model 05730-53, (five 50-ml bottles). All filling solutions for reference electrodes come in cartons of one or five 50-ml bottles.
Specifications
  • Ion-Selective ElectrodeSodium Chloride
  • DescriptionStandardizing solution: NaCl as Cl, 100 ppm
  • Warranty3 Month Product / 12 Month Parts and Labor