Strathkelvin SI782 Precision Dissolved Oxygen Respirometer, two-channel
Representative image only.
Strathkelvin SI782 Precision Dissolved Oxygen Respirometer, two-channel
Strathkelvin | Mfr: SI782| Item: 5300203
Meter interfaces with your PC for respiration rates of mitochondria, cell suspensions, and oxidative enzyme preparations.
Call For Price
Call For Price
Buy It Together
Current Item
Strathkelvin SI782 Precision Dissolved Oxygen Respirometer, two-channel
Strathkelvin SI782 Precision Dissolved Oxygen Respirometer, two-channel
5300203
Call For Price
+ =
Total Price 383000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for laboratory research and cell biology research
  • Precision microcathode electrodes
  • 2 year warranty
More About this Item
Two-channel dissolved oxygen meter operates as a stand-alone meter or as an interface to the computer. The LCD shows dissolved oxygen values: either PO2 (mm Hg, torr, kPa) or concentration (µmol, µg/mL, mg/L, µL/mL, mL/L, percent saturations). Use either one or two electrode input. This meter interfaces to the respirometry software for DO monitoring and closed-cell and flow-cell respirometry.
Six-channel dissolved oxygen meter operates only with respirometry software, with inputs for one to six oxygen electrodes.
The software included with both models has three parts: “setup” to enter experimental parameters and electrode calibration; “recording” to display scrolling traces of oxygen uptake; and “analyze” to recall saved files for automatic calculation of rates from selected parts of the traces.
or a complete system order a respirometer (53002-03 or 53002-21), microcathode electrode (53002-50), and either a Mitocell (53002-26) or Respiration Cell (53002-14). Order electrodes and respiration cells separately.
Not for sales outside North America
Two year warranty
Specifications
  • Meter TypeBenchtop
  • ConnectionsTwo channels: Lemo
  • DescriptionPrecision Dissolved Oxygen Respirometer, two-channel