Strathkelvin FC100 Instruments Oxygen Respirometer Flowcell
Representative image only.
Strathkelvin FC100 Instruments Oxygen Respirometer Flowcell
Strathkelvin | Mfr: FC100| Item: 5300216
Quality accessories for your water analysis
₹159,000.00INR / Each
₹159,000.00INR / Each
Buy It Together
Current Item
Strathkelvin FC100 Instruments Oxygen Respirometer Flowcell
Strathkelvin FC100 Instruments Oxygen Respirometer Flowcell
5300216
₹159,000.00INR / Each
+ =
Total Price 159000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Improve water testing
  • Customize to meet your applications needs
  • Easy to change membranes
More About this Item
Choose from an array of items to improve your water testing requirements.
Flowcell is for monitoring or connecting to a flow-through respirometer chamber. Flowcell features an acetal electrode holder and acrylic chamber with stainless steel inlet and outlet tubes. Acrylic chamber must be immersed in constant temperature bath.
Specifications
  • MaterialAcrylic, acetal, 316 SS
  • Compatible WithStrathkelvin DO respirometer, 53002-03 or 53002-22
  • DescriptionInstruments Oxygen Respirometer Flowcell