Strathkelvin SI130 Oxygen Electrode, Microcathode
Representative image only.
Strathkelvin SI130 Oxygen Electrode, Microcathode
Strathkelvin | Mfr: SI130| Item: 5300250
Quality accessories for your water analysis
₹510,000.00INR / Each
₹510,000.00INR / Each
Buy It Together
Current Item
Strathkelvin SI130 Oxygen Electrode, Microcathode
Strathkelvin SI130 Oxygen Electrode, Microcathode
5300250
₹510,000.00INR / Each
+ + =
Total Price 685000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Improve water testing
  • Customize to meet your applications needs
  • Easy to change membranes
  • 1 month warranty
More About this Item
Choose from an array of items to improve your water testing requirements.
Microcathode Clark-type electrode is supplied with PP membranes. Electrode jacket contains buffered KCl electrolyte solution. May be used in unstirred media if required. Membrane response time is 18 seconds for 90% of change. For fast responses (six seconds for 90% of change) use FEP membranes 53002-55 with electrode jacket 53002-58 sold separately at right.
1 Month Warranty
Specifications
  • MaterialPP membrane
  • Compatible WithStrathkelvin DO respirometer, 53002-03 or 53002-23
  • DescriptionOxygen Electrode, Microcathode
  • Warranty1 month