Strathkelvin MT200A Mitocell respirometer cell, 0.1 to 1.0 mL volume
Representative image only.
Strathkelvin MT200A Mitocell respirometer cell, 0.1 to 1.0 mL volume
Strathkelvin | Mfr: MT200A| Item: 5300226
Quality accessories for your water analysis
₹992,000.00INR / Each
₹992,000.00INR / Each
Buy It Together
Current Item
Strathkelvin MT200A Mitocell respirometer cell, 0.1 to 1.0 mL volume
Strathkelvin MT200A Mitocell respirometer cell, 0.1 to 1.0 mL volume
5300226
₹992,000.00INR / Each
+ =
Total Price 992000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Improve water testing
  • Customize to meet your applications needs
  • Easy to change membranes
  • 2 year warranty
More About this Item
Choose from an array of items to improve your water testing requirements.
Respirometer cells have a glass water jacket for connecting to your pumped source of constant temperature water.
Mitocell miniature respirometer cell is used for mitochondria or small volume cell suspensions. Use with 0.3, 0.5, or 1.0 mL sample size. Seal the central glass respiration chamber by inserting a polycarbonate plunger with central capillary; use to inject substrates and inhibitors. Includes fixed-speed magnetic stirrer and spinbars.
Two year warranty
Specifications
  • MaterialGlass/polycarbonate
  • Compatible WithStrathkelvin DO respirometer, 53002-03 or 53002-22
  • DescriptionMitocell respirometer cell, 0.1 to 1.0 mL volume