Strathkelvin RC350 Respiration Cell, 16-mm diameter, 1.0 to 3.0 mL capacity
Representative image only.
Strathkelvin RC350 Respiration Cell, 16-mm diameter, 1.0 to 3.0 mL capacity
Strathkelvin | Mfr: RC350| Item: 5300214
Quality accessories for your water analysis
₹383,000.00INR / Each
₹383,000.00INR / Each
Buy It Together
Current Item
Strathkelvin RC350 Respiration Cell, 16-mm diameter, 1.0 to 3.0 mL capacity
Strathkelvin RC350 Respiration Cell, 16-mm diameter, 1.0 to 3.0 mL capacity
5300214
₹383,000.00INR / Each
+ =
Total Price 383000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Improve water testing
  • Customize to meet your applications needs
  • Easy to change membranes
More About this Item
Choose from an array of items to improve your water testing requirements.
Respirometer cells have a glass water jacket for connecting to your pumped source of constant temperature water.
Respiration Cell has variable chamber volume. Glass respiration chamber has an acetal electrode holder (included) with a capillary bore for injection of solutions into the cell, using a hypodermic needle (included). Place the cell on a magnetic stirrer. Cells includes PTFE-covered spinbars.
Specifications
  • MaterialGlass/acetal
  • Compatible WithStrathkelvin DO respirometer, 53002-03 or 53002-21
  • DescriptionRespiration Cell, 16-mm diameter, 1.0 to 3.0 mL capacity