Sartorius Arium® Mini Starter Set

Representative image only.

Sartorius Arium® Mini Starter Set

Sartorius | Mfr: H2O-MM-SET-US| Item: 9929446

Starter set includes all the required components to get you up and running

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium® Mini Starter Set
Sartorius Arium® Mini Starter Set
9929446

Call For Price
+
Sartorius H2O-CBS-5-S Arium® Mini Storage Bag, 5 L
9929333

₹15,400.00INR / Each
+ =
Total Price 15400.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Arium Mini system, Scientific kit, 5 L bag, and SterilePlus filter.

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Large touch display with self-explanatory navigation
  • Take up a small footprint—only measures 28 cm in width
  • Volume-controlled dispensing in 50 mL steps
  • Delivers up to 1 L/minute
  • Produces ASTM Type I ultrapure water
  • Stores water in 5 L Bagtank bag
  • 1 year warranty

More About this Item

Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.

Select from Arium Mini, Mini Plus, and Mini Essential depending on your application. All models produce up to 10 L of ASTM Type I ultrapure water per day. Arium Mini and Mini Plus use a closed Bagtank system while Mini Essential has no storage system. For analytical applications, order the Arium Mini with UV lamp to reduce total organic compound (TOC) to = 5 ppb.

Connect Arium Mini water purification system to pretreated water from supply container to produce ASTM Type I ultrapure water. Type I ultrapure water is useful in buffer, media, and reagent preparation and histology, ELISA, AAS, IC and photometry applications.

Arium Mini uses a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals.

Conformite EuropeenneCanadian Standards Association
Specifications
  • ASTM Water GradeType I
  • Capacity5 L
  • Water Purification MethodAdsorption by spherical activated carbon, catalyst, reverse osmosis, ion exchange, and by point-of-use particle-removing filtration
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampNo
  • SourceManual feeed with pretreated water
  • Type I Water (Resistivity)18.2 MΩ × cm
  • Feed Water Requirements (Conductivity)< 100 μS/cm
  • Feed Water Requirements (TOC)< 50 ppb
  • Feed Water Requirements (turbidity)< 1 NTU
  • Type I Water (Conductivity)0.055 μS/cm
  • Type I Water (Bacteria)< 0.01 CFU/mL
  • Feed Water Requirements (pH range)4 to 10
  • Type I Water (Endotoxins)< 0.001 EU/mL
  • Feed Water Requirements (Temperature)2 to 30°C
  • Power (VAC)100 to 240
  • Height (in)20
  • Depth (in)20.9
  • Width (in)11
  • Height (cm)50.9
  • Depth (cm)53.1
  • Width (cm)28
  • DescriptionArium® Mini Starter Set
  • Warranty2 years
Reviews
Q&A