Sartorius Arium Mini Plus Water Purification System with UV lamp

Representative image only.

Sartorius Arium Mini Plus Water Purification System with UV lamp

Sartorius | Mfr: H2O-MA-UV-T-US| Item: 9929322

Eliminate tank cleaning with unique closed bag system

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium Mini Plus Water Purification System with UV lamp
Sartorius Arium Mini Plus Water Purification System with UV lamp
9929322

Call For Price
+
Sartorius H2O-CPR Arium® Mini Plus Pretreatment Cartridge
9929330

₹153,000.00INR / Each
+ =
Total Price 153000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Multicolor touch display with clear icons simplifies operation
  • Compact design with only a 11" (28 cm) wide footprint
  • 2 year warranty

More About this Item

The compact ultrapure water system features a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals. System features include a favorites function to dispense recurring volumes, integrated pump for increasing water pressure, and pretreatment cartridge for effective purification.

Two models are available. The arium mini version is manually fed with pretreated water, while the arium mini plus version has direct connection to the portable water supply. Both are available with UV lamp (185/254 nm) to make sure the water is virtually free of organic components.

Consumables are needed to operate the unit and are sold separately. Please purchase a starter set along with your water system.

Canadian Standards AssociationTwo year warranty
Specifications
  • ASTM Water GradeType I & Type III
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampYes
  • With UltrafilterYes
  • SourceDirect conection to portable water supply
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Type I Water (TOC)≤5 ppb
  • Feed Water Requirements (TOC)<2000 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Type I Water (Bacteria)<1 CFU/1,000 mL
  • Feed Water Requirements (pressure)0.5 to 6 bar
  • Feed Water Requirements (Temperature)2 to 30°C
  • Max Inlet Water Temperature (° C)30
  • Min Inlet Water Temperature (° C)2
  • Max Inlet Pressure (PSI)6
  • Min Inlet Pressure (bar)0.5
  • Power (VAC)100 to 240
  • Height (in)20.06
  • Depth (in)20.89
  • Width (in)11.02
  • Height (cm)50.94
  • Depth (cm)53.07
  • Width (cm)28
  • DescriptionWater Purification System with UV lamp
  • Warranty2 year
Reviews
Q&A