QLA Suppository Basket for Distek; Plastic
Representative image only.
QLA Suppository Basket for Distek; Plastic
| Mfr: BSKSUP-DK| Item: 4130194
QLA Baskets for dissolution testing with your Distek apparatus
₹55,500.00INR / Each
₹55,500.00INR / Each
Buy It Together
Current Item
QLA Suppository Basket for Distek; Plastic
QLA Suppository Basket for Distek; Plastic
4130194
₹55,500.00INR / Each
+
QLA Basket Insertion and Removal Tool for Thick Rim Baskets
4130200
₹146,000.00INR / Each
=
Total Price 201500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of electropolished 316 SS All baskets are laser marked with unique serial numbers
  • Standard push-on style suppository basket
More About this Item
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All (except plastic suppository baskets) are crafted from electropolished 316 stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificate of Conformance for compliance.
.
Specifications
  • MaterialPlastic
  • DescriptionStandard Push-On Style Suppository Basket for Distek, Plastic; Serialized