QLA Push-On Style Standard Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
Representative image only.
QLA Push-On Style Standard Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
| Mfr: BSDRNG-16| Item: 4130121
QLA Shafts for dissolution testing with your Distek apparatus
₹48,700.00INR / Each
₹48,700.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • Standard push-on style
  • 16.5"L
More About this Item
These shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness, and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance.
.
Specifications
  • Material316 stainless steel
  • Compatible WithPush-on style
  • Length (InchShort)16 1/2
  • Length (cm)41.9
  • DescriptionBasket Shaft Standard Push-On Style for Distek 2100, 16.5", 316 SS; Serialized