QLA Distek-Compatible Shafts
QLA Shafts for dissolution testing with your Distek apparatus
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
These shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness, and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance.
48,700.00 - ₹93,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (InchShort) Material Compatible With
Availability Pricing
QLA Push-On Style Standard Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized 4130121 Mfr # BSDRNG-16
COMPARE
16 1/2 316 stainless steel Push-on style
₹48,700.00INR / Each
QLA Push-On Style Standard Basket Shaft for Distek, 19"L, 316 SS; Serialized 4130122 Mfr # BSDRNG-19
COMPARE
19 316 stainless steel Push-on style
₹52,300.00INR / Each
QLA Basket Shaft Assembly for Distek, 16"L, O-Ring, PTFE-Coated 316 SS; Serialized 4130123 Mfr # BSDRNG-TEF16
COMPARE
16 PTFE-coated 316 SS Push-on style
₹93,300.00INR / Each
QLA Spring Clip Style Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized 4130182 Mfr # BSKSHT-16.5
COMPARE
16 1/2 316 stainless steel Evolution style
₹60,300.00INR / Each