QLA Spring Clip Style Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
Representative image only.
QLA Spring Clip Style Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
| Mfr: BSKSHT-16.5 | Item: 4130182
QLA Shafts for dissolution testing with your Distek apparatus
₹60,300.00INR / Each
₹60,300.00INR / Each
Buy It Together
Current Item
QLA Spring Clip Style Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
QLA Spring Clip Style Basket Shaft for Distek 2100, 16.5"L, 316 SS; Serialized
4130182
₹60,300.00INR / Each
+
QLA Basket Clip for Distek Evolution Style Hub; 316 SS
4130168
₹13,400.00INR / Each
=
Total Price 73700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • Spring clip style
  • 16.5"L
More About this Item
These shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness, and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance.
.
Specifications
  • Material316 stainless steel
  • Compatible WithEvolution style
  • Length (InchShort)16 1/2
  • Length (cm)41.9
  • DescriptionSpring Clip Style Basket Shaft for Distek 2100, 16.5", 316 SS; Serialized