QLA Basket Shaft Assembly for Distek, 16"L, O-Ring, PTFE-Coated 316 SS; Serialized
Representative image only.
QLA Basket Shaft Assembly for Distek, 16"L, O-Ring, PTFE-Coated 316 SS; Serialized
| Mfr: BSDRNG-TEF16| Item: 4130123
QLA Shafts for dissolution testing with your Distek apparatus
₹93,300.00INR / Each
₹93,300.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • PTFE-coated 316 SS
  • 16"L
More About this Item
These shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness, and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance.
.
Specifications
  • MaterialPTFE-coated 316 SS
  • Compatible WithPush-on style
  • Length (InchShort)16
  • Length (cm)40.6
  • DescriptionBasket Shaft Assembly, 16", O-Ring, PTFE-Coated 316 SS; Serialized