QLA Distek-Compatible Baskets
QLA Baskets for dissolution testing with your Distek apparatus
  • Made of electropolished 316 SS All baskets are laser marked with unique serial numbers
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All (except plastic suppository baskets) are crafted from electropolished 316 stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificate of Conformance for compliance.
34,700.00 - ₹175,000.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Standard Push-On Style Basket for Distek, 10 Mesh, 316 SS; Serialized 4130129 Mfr # BSK010-DK
COMPARE
316 stainless steel 10 Standard Push-On Style Basket for Distek, 10 Mesh, 316 SS; Serialized
₹41,200.00INR / Each
QLA Evolution Style Basket for Distek, 10 Mesh, 316 SS; Serialized 4130130 Mfr # BSK010-DKC
COMPARE
316 stainless steel 10 Evolution Style Basket for Distek, 10 Mesh, 316 SS; Serialized
₹45,200.00INR / Each
QLA Standard Push-On Style Basket for Distek, 20 Mesh, 316 SS; Serialized 4130137 Mfr # BSK020-DK
COMPARE
316 stainless steel 20 Standard Push-On Style Basket for Distek, 20 Mesh, 316 SS; Serialized
₹43,200.00INR / Each
QLA Evolution Style Basket for Distek, 20 Mesh, 316 SS; Serialized 4130138 Mfr # BSK020-DKC
COMPARE
316 stainless steel 20 Evolution Style Basket for Distek, 20 Mesh, 316 SS; Serialized
₹47,000.00INR / Each
QLA Evolution Style Basket for Distek, 30 Mesh, 316 SS; Serialized 4130144 Mfr # BSK030-DKC
COMPARE
316 stainless steel 30 Evolution Style Basket for Distek, 30 Mesh, 316 SS; Serialized
₹58,700.00INR / Each
QLA Standard Push-On Style Basket for Distek, 40 Mesh, 316 SS; Serialized 4130147 Mfr # BSK040-DK
COMPARE
316 stainless steel 40 Standard Push-On Style Basket for Distek, 40 Mesh, 316 SS; Serialized
₹34,700.00INR / Each
QLA Evolution Style Basket for Distek, 40 Mesh USP, 316 SS; Serialized 4130148 Mfr # BSK040-DKC
COMPARE
316 stainless steel 40 Evolution Style Basket for Distek, 40 Mesh USP, 316 SS; Serialized
₹38,300.00INR / Each
QLA Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized 4130155 Mfr # BSK060-DKC
COMPARE
316 stainless steel 60 Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized
₹68,400.00INR / Each
QLA Evolution Style Basket for Distek, 80 Mesh, 316 SS; Serialized 4130156 Mfr # BSK080-DKC
COMPARE
316 stainless steel 80 Evolution Style Basket for Distek, 80 Mesh, 316 SS; Serialized
₹91,600.00INR / Each
QLA Standard Push-On Style Basket for Distek, 100 Mesh, 316 SS; Serialized 4130159 Mfr # BSK100-DK
COMPARE
316 stainless steel 100 Standard Push-On Style Basket for Distek, 100 Mesh, 316 SS; Serialized
₹82,500.00INR / Each
QLA Evolution Style Basket for Distek, 100 Mesh, 316 SS; Serialized 4130160 Mfr # BSK100-DKC
COMPARE
316 stainless steel 100 Evolution Style Basket for Distek, 100 Mesh, 316 SS; Serialized
₹82,500.00INR / Each
QLA Standard Push-On Style Basket for Distek, 40 Mesh, PTFE Coated over 316 SS; Serialized 4130171 Mfr # BSKPTD-40
COMPARE
PTFE-coated 316 SS 40 Standard Push-On Style Basket for Distek, 40 Mesh, PTFE Coated over 316 SS; Serialized
₹83,900.00INR / Each
QLA Suppository Basket for Distek; Plastic 4130194 Mfr # BSKSUP-DK
COMPARE
Plastic Standard Push-On Style Suppository Basket for Distek, Plastic; Serialized
₹55,500.00INR / Each
QLA Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130840 Mfr # CUSBSK-20/100DK
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Evolution Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130841 Mfr # CUSBSK-20/100DKC
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Evolution Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Standard Push-On Style Basket for Distek, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130847 Mfr # CUSBSK-20/150DK
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Standard Push-On Style Basket for Distek, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Evolution Style Basket for Distek, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130848 Mfr # CUSBSK-20/150DKC
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Evolution Style Basket for Distek, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
Showing 1- 17 of 17