QLA Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
Representative image only.
QLA Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
| Mfr: CUSBSK-20/100DK| Item: 4130840
QLA Baskets for dissolution testing with your Distek apparatus
₹175,000.00INR / Each
₹175,000.00INR / Each
Buy It Together
Current Item
QLA Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
QLA Standard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
4130840
₹175,000.00INR / Each
+
QLA Basket Insertion and Removal Tool for Thick Rim Baskets
4130200
₹146,000.00INR / Each
=
Total Price 321000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of electropolished 316 SS All baskets are laser marked with unique serial numbers
  • Standard push-on style basket
  • 100 mesh with 20 mesh stability lining
More About this Item
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All (except plastic suppository baskets) are crafted from electropolished 316 stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificate of Conformance for compliance.
.
Specifications
  • Material316 stainless steel
  • Mesh Size100 (plus 20 mesh stability lining)
  • DescriptionStandard Push-On Style Basket for Distek, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized