QLA Basket Insertion and Removal Tool for Thick Rim Baskets
Representative image only.
QLA Basket Insertion and Removal Tool for Thick Rim Baskets
| Mfr: BSKTOOL-DK| Item: 4130200
QLA accessories help provide more precise and consistent dissolution results
₹146,000.00INR / Each
₹146,000.00INR / Each
Product Description
Key Features
  • Use to hold dissolution baskets without touching them
More About this Item
Basket tool for picking up and moving dissolution baskets without touching the mesh or rim. Can be used with any standard size basket without damage. Makes it easy to fit basket onto shaft.
.
Specifications
  • Compatible WithThick rim baskets
  • DescriptionBasket Insertion and Removal Tool for Thick Rim Baskets