QLA Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized
Representative image only.
QLA Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized
| Mfr: BSK060-DKC| Item: 4130155
QLA Baskets for dissolution testing with your Distek apparatus
₹68,400.00INR / Each
₹68,400.00INR / Each
Buy It Together
Current Item
QLA Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized
QLA Evolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized
4130155
₹68,400.00INR / Each
+
QLA Basket Clip for Distek Evolution Style Hub; 316 SS
4130168
₹13,400.00INR / Each
=
Total Price 81800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of electropolished 316 SS All baskets are laser marked with unique serial numbers
  • Evolution style basket
  • 60 mesh
More About this Item
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Distek test apparatus. All (except plastic suppository baskets) are crafted from electropolished 316 stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificate of Conformance for compliance.
.
Specifications
  • Material316 stainless steel
  • Mesh Size60
  • DescriptionEvolution Style Basket for Distek, 60 Mesh, 316 SS; Serialized