Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 120V

Representative image only.

Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 120V

Ohaus | Mfr: 83042046| Item: 8524006

Powerful and versatile Multi-Pro centrifuges handles virtually any lab application

₹1,323,000.00INR / Each

₹1,323,000.00INR / Each

Buy It Together

Current Item

Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 120V
Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 120V
8524006

₹1,323,000.00INR / Each
+ + + + + + =
Total Price 4770600.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Rotor key and AC power cable.

Key Features

  • High-speed performance at maximum volume
  • Automatic rotor recognition minimize errors
  • Easy-to-turn rotary knob for quick parameter adjustments
  • Large LCD with simple interface
  • Easily switch between rpm and RCF
  • 10 acceleration/deceleration settings
  • 99 memory sets to quickly store and recall commonly used parameters
  • Preset run time from 10 seconds to 99 hours 59 minutes, continuous, or quick spin
  • Motorized lid-locking system prevents the lid from opening during operation
  • Imbalance sensor stops the rotor when imbalance is detected
  • Refrigerate samples down to -20 °C (-4 °F)
  • High-speed centrifugation with speeds up to 18,000 rpm
  • Accommodates 0.2 to 100 mL test tubes and microplates
  • CFC-free refrigeration system
  • Small footprint with compressor in the rear
  • 2 year warranty

More About this Item

These multipurpose centrifuges feature high-speed performance, automatic rotor recognition, excellent safety features, and accommodate a broad range of rotors and adapters to provide maximum flexibility. Use this powerful centrifuge for high-speed centrifugation even at maximum volume. The automatic rotor recognition feature conveniently stores all rotor specific data, such as maximum speed and acceleration, saving you time and minimizing errors.

Quickly make parameter adjustments using an easy-to-turn rotary knob. Select from 10 acceleration and 10 deceleration rates to suit your application. Store up to 99 runs with relevant parameters, including the used rotors, to easily repeat the procedures and save time. Safety features include a motorized lid-locking system that prevents the lid from opening during operation and an imbalance sensor that stops the rotor when imbalanced load is detected.

Use the refrigerated model to maintain cool sample temperatures. The centrifuge can maintain temperatures from -20 to 40 °C (-4 to 104 °F) ensuring the integrity of our samples; precool feature quickly lowers the temperature of rotor and chamber before a run. E Easily toggle between RPM/RCF, accelerate/decelerate with a single touch. Maintenance-free induction motor produces a maximum RCF of 23,542 x g and reaches speeds up to 18,000 rpm. Chemical- and corrosion-resistant stainless steel housing provides excellent durability. Use sealed and bio-containment rotors to ensure worker safety when handling hazardous materials.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Conformite EuropeenneTwo year warranty
Specifications
  • Max Speed (rpm)18000
  • Min Speed (rpm)200
  • Max RCF x g23542
  • Min RCF x g20
  • Variable SpeedYes
  • Speed ControlDigital, 10 rpm/set
  • Max Temperature (° C)40
  • Min Temperature (° C)-20
  • Max Temperature (° F)104
  • Min Temperature (° F)-4
  • Temp ControlDigital
  • DisplayTwo-line, backlit LCD
  • RefrigeratedYes
  • Motor TypeInduction
  • Noise level (dB(A))≤60
  • Timer Range10 sec to 99 hr 99 min 59 sec, continuous, or quick spin
  • Max Capacity (mL)200
  • Tube Capacity4
  • Length (in)28 13/16
  • Width (in)16 1/8
  • Height (in)14 1/4
  • Length (cm)73
  • Width (cm)40.7
  • Height (cm)36
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (amps)6
  • DescriptionMulti-Pro Refrigerated Centrifuge, 18,000 rpm, 120V
  • Warranty2 years
Reviews
Q&A