InnoCal NIST-Traceable Calibration: Balance/Scale - All Types
Representative image only.
InnoCal NIST-Traceable Calibration: Balance/Scale - All Types
InnoCal | Item: 1710152
NIST-traceable calibrations for balance and scales of all types
₹27,400.00INR / Each
for an additional
₹46,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹27,400.00INR / Each
Customers Also Bought
Product Description
Includes
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI u
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Calibration for balances and scales
  • 1 year warranty
1 Year Warranty
Specifications
  • Calibration TypeForce and Torque
  • Instrument Only CalibrationYes
  • Number Of Calibration Test Points10 points plus shift and repeatibility
  • Calibration Test PointsAcross the range of the instrument
  • DescriptionNIST-Traceable Calibration: Balances/Scales - All Types