Cole-Parmer® LB-200 Analytical Balances
Performance and style for an impressive weighing experience
  • Selectable digital filtering for dynamic weighing enables consistent results for moving subjects
  • Printouts include date and time for data tracking within Good Laboratory Practices (GLP) guidelines
  • Sealed keypad protects against dirt and spills
  • Hold function freezes the displayed weight ensuring results are kept safe
  • Parts counting with preset sample sizes
  • Auto power-off to save energy
Designed to deliver improved performance, the LB-200 Analytical Balances enable faster and more accurate readings, saving valuable time while maintaining precision in measurements. Backlighting ensures you can easily read measurement results in all lighting conditions to eliminate potential sources of error due to poor visibility. Multilingual support makes it adaptable for use in various regions.
Color-coded navigation buttons allow you perform functions with minimal training. USB and RS-232 interfaces facilitate seamless data collection and transmission to computers or printers.
327,000.00 - ₹404,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
COMPARE
External 120 0.0001
₹327,000.00INR / Each
COMPARE
External 220 0.0001
₹349,000.00INR / Each
COMPARE
Internal or External 120 0.0001
₹390,000.00INR / Each
COMPARE
Internal or External 220 0.0001
₹404,000.00INR / Each