Traceable® Digital Thermometers with Calibration
Monitor minimum and maximum temperatures— all at a single glance!
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Triple-display—shows high, low, and current temperature simultaneously
Easily monitor temperature in liquids, baths, air/gas, incubators, refrigerators, or freezers. Keep track of temperature conditions overnight or for any time period—minimum and maximum temperatures are stored until the memory is cleared. Models feature a triple display that simultaneously shows current, minimum, and maximum temperatures. Set high and low alarms in 1° increments; alarm sounds when temperature rises above or falls below set points, and continues to sound even if temperature returns to nonalarm range. Thermometer reads ambient temperature or probe temperature. Probe is attached to a 10-ft (3 m) micro-cable that permits a doors to close on it; probe and cable are both submersible. Mount meter with included hook-and-loop strips, wall-mount slots, or use flip-open stand on benchtop.
7,400.00 - ₹15,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
158 -58 Calibrated General-Purpose Digital Thermometer; 1 Bullet Probe
₹11,000.00INR / Each
COMPARE
158 -58 Calibrated Fridge/Freezer Digital Thermometer; 5 mL Bottle Probe
₹7,400.00INR / Each
COMPARE
158 -58 Calibrated Fridge/Freezer Digital Thermometer; 1 Bottle Probe
₹15,000.00INR / Each