Cole-Parmer® LB-200 Toploading Balances
Performance and style for an impressive weighing experience
  • Level indicator and adjustable feet ensure proper balance setup for optimum weighing results
  • Sealed keypad protects against dirt and spills
  • Hold function freezes the displayed weight, allowing time to elapse without losing the result
  • Parts counting with preset sample sizes
  • Printouts include date and time for data tracking within Good Laboratory Practices (GLP) guidelines
  • Selectable digital filtering for animal/dynamic weighing enables consistent results for moving subjects
  • Auto power-off to save energy
LB-200 toploading balances are designed with an array of features and functions that accommodate most lab applications. This range offers improved performance for faster, more accurate readings. With stylish, modern backlighting, the impressive display is easily visible in any lighting condition. The LCD features large, readily discernible 24 mm digits. Multilingual text and display capability make LB-200 well-suited for users in various regions. For simple operation, navigation buttons are color-coded, helping users perform functions with minimal training. LB-200 provides USB and RS-232 interfaces to facilitate data collection and transmission to computers or printers. ABS plastic construction and the rugged grade 304 stainless steel pan are easy to clean and can withstand demanding lab use.
Toploading balances offer a wider range of weighing capacity, from 210 g up to 6000 g.
133,000.00 - ₹303,000.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g)
Availability Pricing
COMPARE
External 210
₹133,000.00INR / Each
COMPARE
External 420
₹159,000.00INR / Each
COMPARE
Internal 210
₹228,000.00INR / Each
COMPARE
Internal 420
₹164,000.00INR / Each
COMPARE
Internal 800
₹193,000.00INR / Each
COMPARE
External 800
₹257,000.00INR / Each
COMPARE
External 2200
₹133,000.00INR / Each
COMPARE
External 4200
₹159,000.00INR / Each
COMPARE
External 6000
₹271,000.00INR / Each
COMPARE
Internal 2200
₹164,000.00INR / Each
COMPARE
Internal 4200
₹189,000.00INR / Each
COMPARE
Internal 6000
₹303,000.00INR / Each
COMPARE
External 6000
₹145,000.00INR / Each
Showing 1- 13 of 13