Idex 1325 Chromatography Tubing Kit, ETFE; 1/EA
Representative image only.
Idex 1325 Chromatography Tubing Kit, ETFE; 1/EA
Idex | Mfr: 1325| Item: 0201327
Withstands the highest pressures of fluoropolymer tubing
₹53,600.00INR / Each
₹53,600.00INR / Each
Buy It Together
Current Item
Idex 1325 Chromatography Tubing Kit, ETFE; 1/EA
Idex 1325 Chromatography Tubing Kit, ETFE; 1/EA
0201327
₹53,600.00INR / Each
+ + +
Cole-Parmer TPE Tubing, Black, 1/8" ID x 1/4" OD; 25 Ft
5012122
₹3,200.00INR / Pkg of 1
+ +
Cole-Parmer PVC Lab Tubing, 1/8" ID x 3/16" OD; 50 Ft
5012115
₹7,500.00INR / Pkg of 1
+ +
Cole-Parmer FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011915
₹13,300.00INR / Pkg of 1
=
Total Price 123900.00 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Two 1/16" OD x 0.030" ID x 5 ft L tubing 02013-50, two 1/16" OD x 0.010" ID x 5 ft L tubing 02013-51, two 1/8" OD x 0.062" ID x 5 ft L tubing 02014-71, and one polymer tubing cutter 02013-29.
Key Features
  • Excellent chemical resistance
  • Max continuous operating temperature of 176°F (80°C)
  • Includes three most common sizes in HPLC systems
  • 1 year warranty
More About this Item
ETFE offers low gas permeability, excellent mechanical stability, and high pressure-holding capabilities. Due to its low gas permeability, ETFE is an ideal choice for connecting the system pump to the vacuum degasser which will help prevent regassing of mobile phase solvents in transit.
Tubing kit provides three of the most common inner diameters of ETFE tubing used in HPLC systems.
1 Year Warranty
Specifications
  • Length (")60
  • Tubing OD (in)1/16
  • Tubing ID (in)1/100
  • ColorNatural
  • Length (ft)5
  • DescriptionChromatography Tubing Kit, ETFE; 1/EA