Idex® Super Flangeless™ Ferrules, 1/4-28 or M6 Flat Bottom
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,200.00 - ₹17,700.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
Idex Super Flangeless™ Ferrule, 2500 psi Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK 0201372 Mfr # P-250X
COMPARE
Natural 1/16 Super Flangeless™ Ferrule, 2500 psi Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK
₹11,700.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK 0201374 Mfr # P-259X
COMPARE
Yellow 1/16 Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK
₹10,600.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, 2500 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA 0202158 Mfr # P-250
COMPARE
Natural 1/16 Super Flangeless™ Ferrule, 2500 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
₹1,300.00INR / Each
Idex Super Flangeless™ Ferrule, 1850 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA 0202169 Mfr # P-260
COMPARE
Natural 1/16 Super Flangeless™ Ferrule, 1850 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
₹2,000.00INR / Each
Idex Super Flangeless™ Ferrule, 1850 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK 0202170 Mfr # P-260X
COMPARE
Natural 1/16 Super Flangeless™ Ferrule, 1850 psi, Natural PEEK/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK
₹17,700.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Black PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA 0202194 Mfr # P-352
COMPARE
Black 1/8 Super Flangeless™ Ferrule, Black PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
₹1,400.00INR / Each
Idex Super Flangeless™ Ferrule, Black PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK 0202195 Mfr # P-352X
COMPARE
Black 1/8 Super Flangeless™ Ferrule, Black PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK
₹12,200.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Neutral PCTFE/SS Ring, 1.8 mm OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA 0202196 Mfr # P-355
COMPARE
Neutral 5/64 Super Flangeless™ Ferrule, Neutral PCTFE/SS Ring, 1.8 mm OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
₹1,900.00INR / Each
Idex Super Flangeless™ Ferrule, Neutral PCTFE/SS Ring, 1.8 mm OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK 0202197 Mfr # P-355X
COMPARE
Neutral 5/64 Super Flangeless™ Ferrule, Neutral PCTFE/SS Ring, 1.8 mm OD Tubing, 1/4-28 or M6 Flat Bottom; 10/PK
₹12,100.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA 0202314 Mfr # P-259
COMPARE
Yellow 1/16 Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/16" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
₹1,200.00INR / Each
Showing 1- 10 of 10