Idex® Super Flangeless™ Nuts, 1/4-28 Flat Bottom for 1/32" or 1/16" OD Tubing
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
490.00 - ₹99,400.00INR / Pkg of 10
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Nut, Standard Knurl, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201352 Mfr # LT-115X
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹10,900.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201371 Mfr # P-232X
COMPARE
Natural PEEK Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹10,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201373 Mfr # P-255X
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹10,900.00INR / Pkg of 10
Idex Super Flangeless™ FlushNut™, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202109 Mfr # F-356
COMPARE
Natural Stainless steel Super Flangeless™ FlushNut™, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹4,400.00INR / Each
Idex Super Flangeless™ FlushNut™, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202110 Mfr # F-356X
COMPARE
Natural Stainless steel Super Flangeless™ FlushNut™, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹42,000.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Hex, Short, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202113 Mfr # LT-105
COMPARE
Natural Stainless steel Super Flangeless™ Nut, Hex, Short, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹9,900.00INR / Each
Idex Super Flangeless™ Nut, Hex, Short, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202114 Mfr # LT-105X
COMPARE
Natural Stainless steel Super Flangeless™ Nut, Hex, Short, 316 SS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹99,400.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202115 Mfr # LT-115
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Blue PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202116 Mfr # LT-115B
COMPARE
Blue PEEK Super Flangeless™ Nut, Standard Knurl, Short, Blue PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹2,300.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202117 Mfr # LT-115BLK
COMPARE
Black PEEK Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Green PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202118 Mfr # LT-115G
COMPARE
Green PEEK Super Flangeless™ Nut, Standard Knurl, Short, Green PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹2,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Red PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202121 Mfr # LT-115R
COMPARE
Red PEEK Super Flangeless™ Nut, Standard Knurl, Short, Red PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,800.00INR / Each
Idex Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202151 Mfr # P-232
COMPARE
Natural PEEK Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202152 Mfr # P-246
COMPARE
Natural PFA Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 25/PK
₹800.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202153 Mfr # P-246X
COMPARE
Natural PFA Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹7,400.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Double Wing, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202159 Mfr # P-251
COMPARE
Natural PEEK Super Flangeless™ Nut, Double Wing, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 25/PK
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Gray POM, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202160 Mfr # P-252
COMPARE
Gray POM Super Flangeless™ Nut, Standard Knurl, Gray POM, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹490.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Gray POM, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202162 Mfr # P-252X
COMPARE
Gray POM Super Flangeless™ Nut, Standard Knurl, Gray POM, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹4,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202163 Mfr # P-255
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202165 Mfr # P-255BLK
COMPARE
Black PEEK Super Flangeless™ Nut, Standard Knurl, Black PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Showing 1 - 20 of 26