Idex Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA
Representative image only.
Idex Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA
Idex | Mfr: A-327| Item: 0201329
Easily make clean cuts with tubing cutters
₹10,100.00INR / Each
₹10,100.00INR / Each
Buy It Together
Current Item
Idex Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA
Idex Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA
0201329
₹10,100.00INR / Each
+ + +
Idex A-328 Replacement Blades for Polymer Tubing Cutters; 5/PK
0201686
₹2,500.00INR / Pkg of 5
+ + =
Total Price 56900.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Five replacement blades.
Key Features
  • Cutters are available for different tubing sizes and materials
  • Use with 1/16" to 1/8" OD polymer tubing
  • 1 month warranty
More About this Item
Choose from tubing cutters for a wide range of tubing sizes, suitable for a wide range of applications.
Standard polymer tubing cutter features guide holes to provide flat, 90°, burr-free cuts on polymer tubing.
1 Month Warranty
Specifications
  • For Tubing OD (in)1/16
  • Max Tubing OD (in)1/8
  • DescriptionPolymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA