Environmental Express T80IR Portable Infrared Light Turbidity Meter Kit
Representative image only.
Environmental Express T80IR Portable Infrared Light Turbidity Meter Kit
Environmental Express | Item: 3563522
Easily measure water sample turbidity in the field or on the bench
₹149,000.00INR / Each
₹149,000.00INR / Each
Buy It Together
Current Item
Environmental Express T80IR Portable Infrared Light Turbidity Meter Kit
Environmental Express T80IR Portable Infrared Light Turbidity Meter Kit
3563522
₹149,000.00INR / Each
+
Mettler Toledo Max Pro-ISM pH Electrode; MultiPin™
3590030
₹239,000.00INR / Each
+ + + + =
Total Price 626700.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Turbidimeter, calibration solutions (0.0, 20.0, 100, 800 NTU), carrying case, microfiber cloth, screwdriver (to remove battery cover), four AA batteries, silicone oil (10 mL), six sample vials, and user manual.
Key Features
  • Rugged, portable design adapts to use in the field or on your lab bench
  • Simplistic user interface for easy operation at any skill level
  • Standards included as a kit so your technicians are ready to calibrate and measure right out of the box!
  • Large TFT color screen for easy operation
  • Improved accuracy with average measurement mode
  • Battery operated and IP67-rated housing—ideal for use in the field
  • Seamless method compliance for with European standard ISO 7027 (DIN EN 27027)
  • Also available in a White light configuration (part number: 35635-21 for compliance with EPA method 180.1)
More About this Item
US EPA Safe drinking water regulations require measurement of turbidity every 15 minutes. Wastewater facilities require turbidity measurement before and after filter bed backflush. In any lab or field setting, Environmental Express turbidity meters provide adaptability needed to easily complete timely, error free testing requirements.
Our portable turbidity meters have a rugged waterproof, IP67-rated design suitable for use in all environments. Automatic functions like “continuous” measurement mode generate maximum, and minimum measured values for multiple readings as well as averaging for ≥10 readings. Automatic accurate measurement and data treatments means testing schedules are easily maintained without disruption.
Two year warrantyConformite Europeenne
Specifications
  • Meter TypeTurbidity
  • Display typeTFT color screen
  • ConnectionsNone
  • CalibrationYes
  • Min Turbidity0 NTU
  • Max Turbidity1000 NTU
  • Turbidity Resolution0.01/0.1/1 NTU (FNU)
  • Turbidity Accuracy≤ ±2 % of reading + stray light
  • WaterproofIP67
  • Battery4 AA (included)
  • Accessories IncludedCalibration solutions (0.0, 20.0, 100, 800 NTU), carrying case, microfiber cloth, screwdriver (to remove battery cover), four AA batteries, silicone oil (10 mL), six sample vials, and user manual.
  • DescriptionPortable Infrared Light Turbidity Meter Kit
  • Warranty2 years