Environmental Express T80 0 NTU Calibration Solution for T80 Turbidity Meters
Representative image only.
Environmental Express T80 0 NTU Calibration Solution for T80 Turbidity Meters
Environmental Express | Item: 3563524
Calibration solutions for use with T-80 model turbidity meters
₹18,100.00INR / Each
₹18,100.00INR / Each
Buy It Together
Current Item
Environmental Express T80 0 NTU Calibration Solution for T80 Turbidity Meters
Environmental Express T80 0 NTU Calibration Solution for T80 Turbidity Meters
3563524
₹18,100.00INR / Each
+ + + =
Total Price 318800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sample Test Cuvettes sold separately
  • Choose the correct NTU value for your needs
  • 100 mL bottle
More About this Item
The calibration solutions kit can only be used with the T-80 infrared turbidity meter. The 0.0 NTU solution is suitable for use with the T-80 infrared and white light meters.
Specifications
  • Capacity (mL)100
  • Description0 NTU Calibration Solution for T80 Turbidity Meters