Environmental Express® Turbidimeter Accessories
Calibration solutions for use with T-80 model turbidity meters
  • Sample Test Cuvettes sold separately
  • Choose the correct NTU value for your needs
The calibration solutions kit can only be used with the T-80 infrared turbidity meter. The 0.0 NTU solution is suitable for use with the T-80 infrared and white light meters.
18,100.00 - ₹41,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Turbidity Solution Kit for Infrared Instrument Calibrations
₹41,700.00INR / Each
COMPARE
0 NTU Calibration Solution for T80 Turbidity Meters
₹18,100.00INR / Each