Environmental Express T 80 Turbidity Solution Kit for Infrared Instrument Calibrations
Representative image only.
Environmental Express T 80 Turbidity Solution Kit for Infrared Instrument Calibrations
Environmental Express | Item: 3563523
Calibration solutions for use with T-80 model turbidity meters
₹41,700.00INR / Each
₹41,700.00INR / Each
Buy It Together
Current Item
Environmental Express T 80 Turbidity Solution Kit for Infrared Instrument Calibrations
Environmental Express T 80 Turbidity Solution Kit for Infrared Instrument Calibrations
3563523
₹41,700.00INR / Each
+ + =
Total Price 323300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
20, 100, 400, and 800 NTU solution
Key Features
  • Sample Test Cuvettes sold separately
  • Choose the correct NTU value for your needs
  • For use with 35635-15 and 35635-22 turbidimeters
More About this Item
The calibration solutions kit can only be used with the T-80 infrared turbidity meter. The 0.0 NTU solution is suitable for use with the T-80 infrared and white light meters.
Specifications
  • DescriptionTurbidity Solution Kit for Infrared Instrument Calibrations