Environmental Express® Turbidimeters
Easily measure water sample turbidity in the field or on the bench
  • Rugged, portable design adapts to use in the field or on your lab bench
  • Simplistic user interface for easy operation at any skill level
  • Standards included as a kit so your technicians are ready to calibrate and measure right out of the box!
  • Large TFT color screen for easy operation
  • Improved accuracy with average measurement mode
  • Battery operated and IP67-rated housing—ideal for use in the field
US EPA Safe drinking water regulations require measurement of turbidity every 15 minutes. Wastewater facilities require turbidity measurement before and after filter bed backflush. In any lab or field setting, Environmental Express turbidity meters provide adaptability needed to easily complete timely, error free testing requirements.
Our portable turbidity meters have a rugged waterproof, IP67-rated design suitable for use in all environments. Automatic functions like “continuous” measurement mode generate maximum, and minimum measured values for multiple readings as well as averaging for ≥10 readings. Automatic accurate measurement and data treatments means testing schedules are easily maintained without disruption.
2 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Description
Availability Pricing
COMPARE
1000 NTU 0 NTU Portable White Light Turbidity Meter Kit
₹121,000.00INR / Each
COMPARE
1000 NTU 0 NTU Portable Infrared Light Turbidity Meter Kit
₹112,000.00INR / Each