Ohaus Guardian 3000 Digital Stirring Hot plates
Ensures safe, accurate stirring and heating of your sample
  • Safety system protects from over heating
  • Precise stirring from 80 to 1600 rpm
  • Easy to read LCD display
  • Chemical resistant housing
  • IP21 ingress protection
  • ISO 9001 registration
The Guardian 3000 stirring hot plates are designed to provide economical heating and mixing capabilities for use in all laboratory and educational settings. Key features combine safety, ease of use, and versatility to ensure reliable performance for everyday applications. The models offer powerful, consistent stirring performance from 80 to 1600 rpm.
The SafetyHeat™, early detection system, uses two independent safety controls to continuously monitor the electronics in real-time and automatically shuts off heating before overtemperature conditions occur.
A bright backlit LCD display features large temperature and speed settings, intuitive icons, and clear status indicators, including a prominent hot top indicator that lights up when the heater is abourt 40 °C. Other features include a green indicator that lights when unit is heating and stirring, probe in use icon, single point calibration icon, PET front panel, rotary knob controls, two built-in support rod holders, smooth chemical-resistant top for easy cleaning, IP21 ingress protection, and rugged metal housing designed for long-term use.
The stirring hot plates also offer you the option to use temperature probes (not included) for precise temperature control of the sample.
144,000.00 - ₹162,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Temperature (° C) Power (VAC)
Availability Pricing
Ohaus Guardian 3000 e-G31HSRDS Digital Stirring Hot Plate, 5.3" dia, Ceramic Coated Stainless Steel; 120 VAC 8524131 Mfr # 30680266
COMPARE
1600 380 120
₹145,000.00INR / Each
Ohaus Guardian 3000 e-G31HSRDS Digital Stirring Hot Plate, 5.3" dia, Ceramic Coated Stainless Steel; 230 VAC 8524132 Mfr # 30680254
COMPARE
1600 380 230
₹144,000.00INR / Each
Ohaus Guardian 3000 e-G31HS07C Digital Stirring Hot Plate, 7" x 7", Ceramic; 120 VAC 8524133 Mfr # 30680267
COMPARE
1600 500 120
₹162,000.00INR / Each
Ohaus Guardian 3000 e-G31HS07C Digital Stirring Hot Plate, 7" x 7", Ceramic; 230 VAC 8524134 Mfr # 30680255
COMPARE
1600 500 230
₹162,000.00INR / Each
Ohaus Guardian 3000 e-G31HS04C Digital Stirring Hot Plate, 4" x 4", Ceramic; 120 VAC 8524135 Mfr # 30680268
COMPARE
1600 500 120
₹145,000.00INR / Each
Ohaus Guardian 3000 e-G31HS04C Digital Stirring Hot Plate, 4" x 4", Ceramic; 230 VAC 8524136 Mfr # 30680256
COMPARE
1600 500 230
₹145,000.00INR / Each