Corning CoolRack® XT PCR96 Cooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrifuge Tubes; Gray
Representative image only.
Corning CoolRack® XT PCR96 Cooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrifuge Tubes; Gray
Corning | Mfr: 432053| Item: 0439253
Cool, freeze, heat, or thaw samples uniformly
₹140,000.00INR / Each
₹140,000.00INR / Each
Buy It Together
Current Item
Corning CoolRack® XT PCR96 Cooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrifuge Tubes; Gray
Corning CoolRack® XT PCR96 Cooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrifuge Tubes; Gray
0439253
₹140,000.00INR / Each
+ + + =
Total Price 185800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
More About this Item
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.
1 Year Warranty
Specifications
  • Tubes Held24
  • ColorGray
  • Capacity24 x 1.5/2.0 mL microcentrfuge tubes, 1 x 96-well PCR plate, or 12 Stripwell™
  • DescriptionCooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrfuge Tubes; Gray