Corning CoolRack® Microcentrifuge Tube and PCR Plate Cooling Blocks
Cool, freeze, heat, or thaw samples uniformly
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.
27,500.00 - ₹235,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Color Tubes Held
Availability Pricing
Corning CoolRack® M6 Cooling Block, 6 x 1.5/2 mL Microcentrifuge Tubes; Gray 0439234 Mfr # 432034
COMPARE
6 x 1.5 or 2 mL microcentifuge tubes Gray 6
₹27,500.00INR / Each
Corning CoolRack® M6 Cooling Block, 6 x 1.5/2 mL Microcentrifuge Tubes; Green 0439235 Mfr # 432035
COMPARE
6 x 1.5 or 2 mL microcentifuge tubes Green 6
₹27,500.00INR / Each
Corning CoolRack® M6 Cooling Block, 6 x 1.5/2 mL Microcentrifuge Tubes; Orange 0439236 Mfr # 432036
COMPARE
6 x 1.5 or 2 mL microcentifuge tubes Orange 6
₹27,500.00INR / Each
Corning CoolRack® M15 Cooling Block, 15 x 1.5/2 mL Microcentrifuge Tubes; Gray 0439237 Mfr # 432037
COMPARE
15 x 1.5 or 2 mL microcentifuge tubes Gray 15
₹66,800.00INR / Each
Corning CoolRack® M15 Cooling Block, 15 x 1.5/2 mL Microcentrifuge Tubes; Green 0439238 Mfr # 432038
COMPARE
15 x 1.5 or 2 mL microcentifuge tubes Green 15
₹66,800.00INR / Each
Corning CoolRack® M15 Cooling Block, 15 x 1.5/2 mL Microcentrifuge Tubes; Orange 0439239 Mfr # 432039
COMPARE
15 x 1.5 or 2 mL microcentifuge tubes Orange 15
₹66,800.00INR / Each
Corning CoolRack® XT M24 Cooling Block, 24 x 1.5/2 mL Microcentrifuge Tubes; Gray 0439240 Mfr # 432040
COMPARE
24 x 1.5 or 2 mL microcentifuge tubes Gray 24
₹117,000.00INR / Each
Corning CoolRack® M30 Cooling Block, 30 x 1.5/2 mL Microcentrifuge Tubes; Gray 0439241 Mfr # 432041
COMPARE
30 x 1.5 or 2 mL microcentifuge tubes Gray 30
₹118,000.00INR / Each
Corning CoolRack® M30 Cooling Block, 30 x 1.5/2 mL Microcentrifuge Tubes; Green 0439242 Mfr # 432042
COMPARE
30 x 1.5 or 2 mL microcentifuge tubes Green 30
₹118,000.00INR / Each
Corning CoolRack® M30 Cooling Block, 30 x 1.5/2 mL Microcentrifuge Tubes; Orange 0439243 Mfr # 432043
COMPARE
30 x 1.5 or 2 mL microcentifuge tubes Orange 30
₹118,000.00INR / Each
Corning CoolRack® M90 Cooling Block, 90 x 1.5/2 mL Microcentrifuge Tubes; Gray 0439244 Mfr # 432044
COMPARE
90 x 1.5 or 2 mL microcentifuge tubes Gray 90
₹177,000.00INR / Each
Corning CoolRack® M96 ID Cooling Block, 96 x 1.5/2 mL Microcentrifuge Tubes, Indexed; Gray 0439245 Mfr # 432045
COMPARE
96 x 1.5 or 2 mL microcentifuge tubes Gray 96
₹235,000.00INR / Each
Corning CoolRack® XT PCR96 Cooling Block, 1 x 96-well PCR Plate, or 12 Stripwells/Microcentrifuge Tubes; Gray 0439253 Mfr # 432053
COMPARE
24 x 1.5/2.0 mL microcentrfuge tubes, 1 x 96-well PCR plate, or 12 Stripwell™ Gray 24
₹140,000.00INR / Each
Corning CoolRack® XT M-PCR Cooling Block, 12 x 1.5/2 mL Microcentrifuge Tubes or 6 Stripwells; Gray 0439254 Mfr # 432054
COMPARE
12 x 1.5/2 mL microcentrifuge tubes or 6 Stripwell™ Gray 12
₹137,000.00INR / Each
Corning CoolRack® XT PCR384 Cooling Block, 1 x 384-well PCR Plate; Gray 0439255 Mfr # 432055
COMPARE
1 x 384-well PCR Plate Gray 1
₹154,000.00INR / Each
Showing 1- 15 of 15