Cole-Parmer® Benchtop Cooler Color-Changing Racks
Color changes to indicate temperature of samples
  • Maintains sample temperature of less than 4 °C (39.2 °F) for up to 4 hours
  • Clear lid for easy sample identification
  • Constructed of rugged polypropylene
  • Filled with an inert gel
Benchtop cooler color-changing racks change from green to yellow as temperature rises to 7 °C (44.6 °F). This provides you a visual indication when your samples need to be returned to the freezer. These racks will hold 4 °C (39.2 °F) temperature for up to 4 hours on the bench. Clear lid cover allows you to easily view and identify samples without removal.
Available in two rack sizes.
11,800.00 - ₹13,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Lowest Set Temperature (° C) Tubes Held Tube Size
Availability Pricing
COMPARE
4 24 0.5, 1.5, 2 mL
₹11,800.00INR / Each
COMPARE
4 96 0.1, 0.2 mL
₹13,000.00INR / Pkg of 2