Cole-Parmer® Benchtop Reversible Cooler Racks
Ensures consistent thermal protection of samples
  • Clear lid for easy sample identification
  • Constructed of impact-resistant polycarbonate
  • Filled with a non-toxic self-insulating gel
Reversible polycarbonate cooler racks feature one side designed to hold 1.5- or 2-mL tubes while the other side holds 0.5 mL tubes. The 4 x 5 grid keeps up to 20 tubes upright and secure. Racks are filled with an inert, non-toxic gel to help maintain a consistent temperature. Translucent lid cover allows you to easily view and identify samples without removal.
15,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Lowest Set Temperature (° C) Tube Size
Availability Pricing
COMPARE
Blue -20 0.5, 1.5, 2 mL
₹15,400.00INR / Each
COMPARE
White 0 0.5, 1.5, 2 mL
₹15,400.00INR / Each