Cooper-Atkins 93230-K EconoTemperature Thermometer Kit; Meter/Lanyard/Wall Bracket/Probe
Representative image only.
Cooper-Atkins 93230-K EconoTemperature Thermometer Kit; Meter/Lanyard/Wall Bracket/Probe
Cooper-Atkins | Mfr: 93230-K| Item: 9002517
Designed to withstand harsh environments found in kitchens or food processing areas
₹76,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹76,000.00INR / Each
Buy It Together
Current Item
Cooper-Atkins 93230-K EconoTemperature Thermometer Kit; Meter/Lanyard/Wall Bracket/Probe
Cooper-Atkins 93230-K EconoTemperature Thermometer Kit; Meter/Lanyard/Wall Bracket/Probe
9002517
₹76,000.00INR / Each
+ + =
Total Price 192200.00 ADD 3 ITEMS TO CART
Product Description
Includes
Needle probe, protective rubber boot, wall mount bracket, lanyard, and three AAA batteries
Key Features
  • Powerful microprocessor deliver speed and reliability
  • 5 year warranty
More About this Item
The EconoTemp thermocouple thermometer is ergonomically designed with a curved back to fit in nicely in the palm of your hand. The meter has a unique built-in memory feature that stores calibration settings, reducing the need for frequent recalibrations. The ABS housing provides impact and water resistance, it also has molded tabs on the side to hold and store most needle probes. Other features include °F/°C/K selectable units and auto power-off to save on battery. Meter accepts any type k thermocouple probe with a standard mini-connector.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
5 Year WarrantyConformite Europeenne
Specifications
  • Length (InchShort)7
  • Height (in)1.2968
  • Width (InchShort)2 3/4
  • DescriptionEconoTemperature Thermometer Kit; Meter/Lanyard/Wall Bracket/Probe
  • Warranty5 years