Cooper-Atkins 50209-K Micro-needle thermocouple type K probe with 0.085" (2.16 mm) dia penetration tip
Representative image only.
Cooper-Atkins 50209-K Micro-needle thermocouple type K probe with 0.085" (2.16 mm) dia penetration tip
Cooper-Atkins | Mfr: 50209-K| Item: 9002514
Optimal choice for accuracy and maximum durability in food service applications
₹36,900.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹36,900.00INR / Each
Buy It Together
Current Item
Cooper-Atkins 50209-K Micro-needle thermocouple type K probe with 0.085" (2.16 mm) dia penetration tip
Cooper-Atkins 50209-K Micro-needle thermocouple type K probe with 0.085" (2.16 mm) dia penetration tip
9002514
₹36,900.00INR / Each
+
Cooper-Atkins 35100-K Aquatuff Waterproof Thermocouple Meter
9002507
₹104,000.00INR / Each
+ =
Total Price 183600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • –100 to 500°F (–73 to 260°C)
  • 1 year warranty
1 Year Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-100
  • Max Temperature (° F)500
  • Min Temperature (° C)-73
  • Max Temperature (° C)260
  • Response Time (Sec)1
  • Connector TypeMini Connector
  • Sheath MaterialPolyurethane outer jacket
  • Handle MaterialStainless steel
  • Sheath Diameter (InchShort)0.085
  • Cable TypeCoiled
  • ApplicationPenetration, Low-Temp Needle
  • DescriptionMicro-needle thermocouple type K probe with 0.085" (2.16 mm) dia penetration tip