InnoCal A2LA ISO17025 Accredited Calibration, Thermocouple Thermometer System Meter+Probe
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration, Thermocouple Thermometer System Meter+Probe
InnoCal | Item: 1750006
Regardless of the type of thermometry probe your application requires, InnoCal can ensure the accuracy of your meter, probe, or system
₹44,700.00INR / Each
₹44,700.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration, Thermocouple Thermometer System Meter+Probe
InnoCal A2LA ISO17025 Accredited Calibration, Thermocouple Thermometer System Meter+Probe
1750006
₹44,700.00INR / Each
+ + +
Traceable Thermistor Mini Thermometer
0840260
₹6,400.00INR / Each
+
Traceable Scientific Thermistor Thermometer with USB Probe
9008009
₹81,700.00INR / Each
+ + +
Traceable High-Precision RTD Thermometer
3780392
₹61,500.00INR / Each
+
Traceable Type K Thermocouple Thermometer with Memory
9121005
₹54,200.00INR / Each
+ =
Total Price 304800.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification
  • EPA, USDA, ISO and GLP/GMPs standards require regular NIST-traceable calibration of all test equipment being used in critical applications
  • A2LA Accredited Calibration. View our scope of accreditation
  • Calibration service for a thermocouple thermometer system (meter+probe)
More About this Item
Regardless of the type of thermometry probe your application requires, InnoCal can ensure the accuracy of your meter, probe, or system!
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025 and MIL-STD-45662A. See the sample certificate under the “More Info” section to see how InnoCal’s certificates provide more information in a concise manner than the rest of the industry.
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory which is technically proficient and committed to quality and customer-focused continuous improvement. Our scope of A2LA accreditation can be found under the “More Info” section.
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Temperature
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 points
  • Calibration Test PointsITS-90 points across the range unless specfied
  • Calibration Test Range-196 to 1300 C
  • DescriptionA2LA ISO17025 Accredited Calibration, Thermocouple Thermometer System Meter+Probe