Cooper-Atkins 50014-K Weighted Griddle-Surface Type K Probe with 2-1/2" Dia Head
Representative image only.
Cooper-Atkins 50014-K Weighted Griddle-Surface Type K Probe with 2-1/2" Dia Head
Cooper-Atkins | Mfr: 50014-K| Item: 9002509
Optimal choice for accuracy and maximum durability in food service applications
₹71,500.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹71,500.00INR / Each
Buy It Together
Current Item
Cooper-Atkins 50014-K Weighted Griddle-Surface Type K Probe with 2-1/2" Dia Head
Cooper-Atkins 50014-K Weighted Griddle-Surface Type K Probe with 2-1/2" Dia Head
9002509
₹71,500.00INR / Each
+ +
Cooper-Atkins 35100-K Aquatuff Waterproof Thermocouple Meter
9002507
₹104,000.00INR / Each
=
Total Price 218200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 500°F (260°C) max
  • 1 year warranty
1 Year Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-40
  • Max Temperature (° F)500
  • Min Temperature (° C)-40
  • Max Temperature (° C)260
  • Response Time (Sec)2
  • Connector TypeMini Connector
  • Sheath MaterialFlexible Stainless Steel Armored Sheath
  • Handle MaterialPhenolic
  • Sheath Diameter (InchShort)2.5
  • Cable TypeSS armor
  • ApplicationSurface, Griddle (Weighted)
  • DescriptionWeighted Griddle-Surface Type K Probe with 2-1/2" Dia Head