Cole-Parmer Variable Area Flowmeter, 316 Stainless Steel, Glass Float 1/8" NPT(F), 46 mL/min. (Air), 0.50 mL/min. (H2O), No Valve
Representative image only.
Cole-Parmer Variable Area Flowmeter, 316 Stainless Steel, Glass Float 1/8" NPT(F), 46 mL/min. (Air), 0.50 mL/min. (H2O), No Valve
Cole-Parmer | Item: 1530941
Ideal for precise low flow rate measurements
₹60,400.00INR / Each
for an additional
₹49,800.00INR / Each
for an additional
₹63,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹60,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Variable Area Flowmeter, 316 Stainless Steel, Glass Float 1/8" NPT(F), 46 mL/min. (Air), 0.50 mL/min. (H2O), No Valve
Cole-Parmer Variable Area Flowmeter, 316 Stainless Steel, Glass Float 1/8" NPT(F), 46 mL/min. (Air), 0.50 mL/min. (H2O), No Valve
1530941
₹60,400.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 500 SCCM
1530880
₹169,000.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 1 SLPM
1530881
₹169,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 20 SLPM
1530908
₹222,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
1708000
₹49,800.00INR / Each
+ + =
Total Price 1080100.00 ADD 7 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Mounting fittings.
Key Features
  • Corrosion-resistant stainless-steel fittings ensure long-term reliability
  • Easy installation and exchange of flow tubes
  • Non-rotating adapter prevents tubes from turning
  • Panel mounting with hex nuts makes installation simple and secure
  • Chemical compatibility
  • 1 year warranty
More About this Item
These single flow tube variable area flowmeters are designed for precise low flow rate measurements of air and water in mL/minute at a given set of pressure and temperature parameters. The Optigrad™ 150 mm universal millimeter scale minimizes parallax and eye fatigue. Ideal for metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, liquid and gas measurement in laboratories, and pilot plants.
The flow meters include standard mounting fittings, side plates, thick protective magnifying front shield and back plate, and stable flow tubes for accurate readings.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
1 Year Warranty
Specifications
  • Accuracy±5% of full scale reading
  • Repeatability±0.25%
  • Number of Channels1
  • Width (InchShort)1 1/4
  • Height (InchShort)9 13/16
  • Max Pressure (psig)200
  • Max Air Flow Rate (mLmin)46
  • Max Water Flow Rate (mLmin)0.5
  • Material Flow TubeBorosilicate glass
  • Material Fitting316 Stainless Steel
  • Material O-RingViton®
  • Material FloatGlass
  • Material HousingWhite acrylic back plate, aluminum side panels
  • ValveNo
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))1/8
  • Fluid TypeWater
  • Gas TypeAir
  • DescriptionVariable Area Flowmeter, 316 Stainless Steel, Glass Float 1/8" NPT(F), 46 mL/min. (Air), 0.50 mL/min. (H2O), No Valve