Cole-Parmer VapLock™ Port Plug, PTFE, 5/16"-24 UNF(M); 1/EA
Representative image only.
Cole-Parmer VapLock™ Port Plug, PTFE, 5/16"-24 UNF(M); 1/EA
Cole-Parmer | Item: 1202051
Use plugs to seal unused ports for a leak-resistant system
₹2,300.00INR / Each
₹2,300.00INR / Each
Product Description
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.
Specifications
  • Fitting MaterialPTFE
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (InchShort)5/16"-24
  • ColorWhite
  • DescriptionPort Plug, 5/16"-24 UNF(M), PTFE; 1/ea