Idex® Quick Connect Threaded Male Luer Adapters
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
630.00 - ₹5,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
Idex Threaded Luer Adapter, Natural ETFE, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA WW-02013-99 Mfr # P-625
COMPARE
Male luer slip Male 1/4-28 Flat Bottom Natural ETFE
₹840.00INR / Each
Idex Threaded Luer Adapter, Red/Black PEEK, 0.040" Bore, Male Luer Lock x Female 1/4-28 Flat Bottom; 1/EA WW-02014-12 Mfr # P-655
COMPARE
Male luer lock Female 1/4-28 Flat Bottom Red/Black PEEK
₹3,000.00INR / Each
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.050" Bore, Male Luer Lock x Female 10-32 Coned; 1/EA WW-02014-13 Mfr # P-656
COMPARE
Male luer lock Female 10-32 Coned Natural/Black PEEK
₹5,000.00INR / Each
Idex Threaded Luer Adapter, Red ETFE/Natural PP, 0.050" Bore, Male Luer Lock x Female 1/4-28 Flat Bottom; 1/EA WW-02014-21 Mfr # P-675
COMPARE
Male luer lock Female 1/4-28 Flat Bottom Red ETFE/Natural PP
₹2,000.00INR / Each
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Luer Lock x Male 1/4-28 Flat Bottom; 1/EA WW-02014-24 Mfr # P-683
COMPARE
Male luer lock Male 1/4-28 Flat Bottom Natural/Black PEEK
₹4,400.00INR / Each
Idex Threaded Luer Adapter, Natural Polypropylene, 0.050" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA WW-02023-40 Mfr # P-619
COMPARE
Male luer slip Male 1/4-28 Flat Bottom Natural Polypropylene
₹630.00INR / Each
Idex Threaded Luer Adapter, Red/Black PEEK, 0.040" Bore, Male Slip Luer x Female 1/4-28 Flat Bottom; 1/EA WW-02023-56 Mfr # P-655-01
COMPARE
Male luer slip Female 1/4-28 Flat Bottom Red/Black PEEK
₹2,500.00INR / Each
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.050" Bore, Male Slip Luer x Female 10-32 Coned; 1/EA WW-02023-57 Mfr # P-656-01
COMPARE
Male luer slip Female 10-32 Coned Natural/Black PEEK
₹2,500.00INR / Each
Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA WW-02023-58 Mfr # P-657
COMPARE
Male luer lock Female M6 Flat Bottom Black PEEK
₹3,000.00INR / Each
Idex Threaded Luer Adapter, Red ETFE/Natural PP, 0.050" Bore, Male Slip Luer x Female 1/4-28 Flat Bottom; 1/EA WW-02023-62 Mfr # P-675-01
COMPARE
Male luer slip Female 1/4-28 Flat Bottom Red ETFE/Natural PP
₹1,600.00INR / Each
Idex Threaded Luer Adapter, Black ETFE/PP, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA WW-02023-63 Mfr # P-677
COMPARE
Male luer lock Female M6 Flat Bottom Black ETFE/PP
₹2,000.00INR / Each
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA WW-02023-66 Mfr # P-683-01
COMPARE
Male luer slip Male 1/4-28 Flat Bottom Natural/Black PEEK
₹4,000.00INR / Each
Showing 1- 12 of 12