Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-683-01| Item: 0202366
Convenience of quick connect and disconnect of a luer connection
₹7,200.00INR / Each
₹7,200.00INR / Each
Buy It Together
Current Item
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA
Idex Threaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA
0202366
₹7,200.00INR / Each
+ + + + + + + + + =
Total Price 40110.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Manufactured from inert polymers
  • Male Slip Luer x Male 1/4-28 Flat Bottom
  • 0.040" thru-hole diameter
  • Manufactured from PEEK
  • 1 year warranty
More About this Item
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
1 Year Warranty
Specifications
  • MaterialNatural/Black PEEK
  • Connection Type-Port 1Male luer slip
  • Connection Type-Port 2Male 1/4-28 Flat Bottom
  • DescriptionThreaded Luer Adapter, Natural/Black PEEK, 0.040" Bore, Male Slip Luer x Male 1/4-28 Flat Bottom; 1/EA