Idex® Flangeless Standard Knurl Fitting Assemblies, 1/4-28 Flat-Bottom Connection for 1/16" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
990.00 - ₹16,700.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flangeless Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201495 Mfr # XP-201X
COMPARE
Black 1/16 Acetal
₹9,800.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202063 Mfr # XP-201
COMPARE
Black 1/16 Acetal
₹990.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Red Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202064 Mfr # XP-202
COMPARE
Red 1/16 Acetal
₹1,100.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Red Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202065 Mfr # XP-202X
COMPARE
Red 1/16 Acetal
₹9,900.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202076 Mfr # XP-230
COMPARE
Natural 1/16 PEEK
₹1,800.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/Pk 0202077 Mfr # XP-230X
COMPARE
Natural 1/16 PEEK
₹16,700.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Purple Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202079 Mfr # XP-238
COMPARE
Purple 1/16 Acetal
₹1,600.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Blue PFA, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202081 Mfr # XP-245B
COMPARE
Blue 1/16 PFA
₹1,500.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PFA, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/Pk 0202082 Mfr # XP-245X
COMPARE
Natural 1/16 PFA
₹12,500.00INR / Pkg of 10
Showing 1- 9 of 9