Idex Flangeless Fitting, Standard Knurl, Red Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Fitting, Standard Knurl, Red Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Idex | Mfr: XP-202X| Item: 0202065
Eliminate the need to flange tubing
₹9,900.00INR / Pkg of 10
₹9,900.00INR / Pkg of 10
Buy It Together
Current Item
Customers Also Bought
Product Description
Includes
Blue ETFE ferrules 02019-33.
Key Features
  • Removable and reusable—saving you money!
  • Red acetal
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorRed