IDEX® Natural FEP Chromatography Tubing
Excellent chemical resistance at an economical price
  • Great for moderate to low pressure applications
  • Low gas permeability
FEP (fluorinated ethylene propylene) tubing is ideal choice for general, low pressure applications. Most FEP tubing is translucent, so you can see liquid flow.
5,700.00 - ₹173,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
Idex 1520-5 Chromatography Tubing, Natural FEP, 1/16" OD x 0.030" ID x 5 ft L; 1/EA 0201296 Mfr # 1520-5
COMPARE
5 1/32 1/16
₹8,400.00INR / Each
Idex 1521-5 Chromatography Tubing, Natural FEP, 1/8" OD x 0.062" ID x 5 ft L; 1/EA 0201297 Mfr # 1521-5
COMPARE
5 1/16 1/8
₹8,800.00INR / Each
Idex 1524XL Chromatography Tubing, Natural FEP, 3/16" OD x 0.125" ID x 100 ft L; 1 /EA 0201298 Mfr # 1524XL
COMPARE
100 1/8 3/16
₹107,000.00INR / Each
Idex 1548L Chromatography Tubing, Natural FEP, 1/16" OD x 0.020" ID x 50 ft L; 1/EA 0201304 Mfr # 1548L
COMPARE
50 1/50 1/16
₹34,700.00INR / Each
Idex 1520L Chromatography Tubing, Natural FEP, 1/16" OD x 0.030" ID x 50 ft L; 1/EA 0201473 Mfr # 1520L
COMPARE
50 1/32 1/16
₹34,900.00INR / Each
Idex 1520XL Chromatography Tubing, Natural FEP, 1/16" OD x 0.030" ID x 100 ft L; 1/EA 0201476 Mfr # 1520XL
COMPARE
100 1/32 1/16
₹58,600.00INR / Each
Idex 1521L Chromatography Tubing, Natural FEP, 1/8" OD x 0.062" ID x 50 ft L; 1/EA 0201477 Mfr # 1521L
COMPARE
50 1/16 1/8
₹51,700.00INR / Each
Idex 1521XL Chromatography Tubing, Natural FEP, 1/8" OD x 0.062" ID x 100 ft L; 1/EA 0201478 Mfr # 1521XL
COMPARE
100 1/16 1/8
₹89,100.00INR / Each
Idex 1524L Chromatography Tubing, Natural FEP, 3/16" OD x 0.125" ID x 50 ft L; 1/EA 0201479 Mfr # 1524L
COMPARE
50 1/8 3/16
₹63,700.00INR / Each
Idex 1527-5 Chromatography Tubing, Natural FEP, 1/16" OD x 0.010" ID x 5 ft L; 1/EA 0201486 Mfr # 1527-5
COMPARE
5 1/100 1/16
₹6,500.00INR / Each
Idex 1527L Chromatography Tubing, Natural FEP, 1/16" OD x 0.010" ID x 50 ft L; 1/EA 0201505 Mfr # 1527L
COMPARE
50 1/100 1/16
₹40,000.00INR / Each
Idex 1527XL Chromatography Tubing, Natural FEP, 1/16" OD x 0.010" ID x 100 ft L; 1/EA 0201507 Mfr # 1527XL
COMPARE
100 1/100 1/16
₹69,500.00INR / Each
Idex 1548-5 Chromatography Tubing, Natural FEP, 1/16" OD x 0.020" ID x 5 ft L; 1/EA 0201508 Mfr # 1548-5
COMPARE
5 1/50 1/16
₹5,700.00INR / Each
Idex 1548XL Chromatography Tubing, Natural FEP, 1/16" OD x 0.020" ID x 100 ft L; 1/EA 0201512 Mfr # 1548XL
COMPARE
100 1/50 1/16
₹59,100.00INR / Each
Idex 1650L Chromatography Tubing, Natural FEP, 1/4" OD x 0.188" ID x 50 ft L; 1/EA 0201513 Mfr # 1650L
COMPARE
50 3/16 1/4
₹102,000.00INR / Each
Idex 1650XL Chromatography Tubing, Natural FEP, 1/4" OD x 0.188" ID x 100 ft L; 1/EA 0201514 Mfr # 1650XL
COMPARE
100 3/16 1/4
₹173,000.00INR / Each
Idex 1671L Chromatography Tubing, Natural FEP, 0.040" OD x 0.020" ID x 50 ft L; 1/EA 0201515 Mfr # 1671L
COMPARE
50 1/50 1/25
₹52,600.00INR / Each
Idex 1673L Chromatography Tubing, Natural FEP, 0.080" OD x 0.040" ID x 50 ft L; 1/EA 0201516 Mfr # 1673L
COMPARE
50 1/25 5/64
₹52,600.00INR / Each
Idex 1677L Chromatography Tubing, Natural FEP, 0.120" OD x 0.080" ID x 50 ft L; 1/EA 0201517 Mfr # 1677L
COMPARE
50 5/64 1/8
₹52,600.00INR / Each
Showing 1- 19 of 19