Idex® Flangeless Standard Knurl Fitting Assemblies, 1/4-28 Flat-Bottom Connection for 1/8" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
620.00 - ₹9,600.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flangeless Fitting, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK WW-02014-99 Mfr # XP-301X
COMPARE
Black 1/8 Acetal
₹5,600.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK WW-02015-03 Mfr # XP-330X
COMPARE
Natural 1/8 PEEK
₹9,600.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02020-85 Mfr # XP-301
COMPARE
Black 1/8 Acetal
₹900.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Green Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02020-90 Mfr # XP-305
COMPARE
Green 1/8 Acetal
₹900.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Green Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK WW-02020-91 Mfr # XP-305X
COMPARE
Green 1/8 Acetal
₹5,600.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02020-94 Mfr # XP-315
COMPARE
Natural 1/8 ETFE
₹780.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/Pk WW-02020-95 Mfr # XP-315X
COMPARE
Natural 1/8 ETFE
₹7,000.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural PP, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02020-97 Mfr # XP-320
COMPARE
Natural 1/8 PP
₹620.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02020-98 Mfr # XP-330
COMPARE
Natural 1/8 PEEK
₹1,400.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA WW-02021-01 Mfr # XP-345
COMPARE
Natural 1/8 PFA
₹800.00INR / Each
Showing 1- 10 of 10