Idex Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: XP-315| Item: 0202094
Eliminate the need to flange tubing
₹1,400.00INR / Each
₹1,400.00INR / Each
Buy It Together
Current Item
Customers Also Bought
Product Description
Includes
Yellow ETFE ferrule 02019-82.
Key Features
  • Removable and reusable—saving you money!
  • Natural ETFE
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8